
1,10-Phenanthroline-2,9-dicarboxylic acid hydrate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:57709-61-2
:
Formula:C₁₄H₈N₂O₄·xH₂O
MDL Number:MFCD00209670
:268.23 g/mol (anhy)
:GHS07
Storage Temperature:Ambient