

2,3-Dibromopropionyl chloride 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:18791-02-1
:
Formula:C₃H₃Br₂ClO
MDL Number:MFCD00000712
:250.31 g/mol
Boiling Pt:191…193 °C
Density:2.181
Flash Pt:67 °C (152.6 °F)
Storage Temperature:Ambient
EINECS:242-575-2
ADR:8,II
:GHS05
:GHS07