Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
EDTA tetrasodium salt hydrate
EDTA tetrasodium salt hydrate 98%
EDTA tetrasodium salt hydrate 98%
:
Thermo Fisher Scientific
EDTA tetrasodium salt hydrate 98%
:
Thermo Fisher Scientific
:
194491-31-1
:
C₁₀H₁₂N₂O₈Na₄·xH₂O
:
MFCD00150025
:
380.17 g/mol (anhy)
:
>300 °C
:
200-573-9
:
GHS05
:
GHS07
:
Ambient
msds_ALFAA17385
Thermo Scientific Acroseal 25 Years