

1H-Pyrazolo[3,4-d]pyrimidin-4(5H)-one 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:315-30-0
:
Formula:C₅H₄N₄O
MDL Number:MFCD00599413
:136.11 g/mol
Melting Pt:>300 °C
EINECS:206-250-9
ADR:6.1,III
:GHS06
:GHS07
Storage Temperature:Ambient