Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
1,6-Anhydro-beta-D-glucose
1,6-Anhydro-β-D-glucose 99%
1,6-Anhydro-β-D-glucose 99%
:
Thermo Fisher Scientific
1,6-Anhydro-β-D-glucose 99%
:
Thermo Fisher Scientific
:
498-07-7
:
:
C₆H₁₀O₅
:
MFCD00063248
:
162.14 g/mol
:
180…184 °C
:
Ambient
:
207-855-0
msds_ALFAA16724
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years