Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Chloropropylamine hydrochloride
3-Chloropropylamine hydrochloride 98%
3-Chloropropylamine hydrochloride 98%
:
Thermo Fisher Scientific
3-Chloropropylamine hydrochloride 98%
:
Thermo Fisher Scientific
:
6276-54-6
:
:
C₃H₈ClN
:
MFCD00012913
:
130.01 g/mol
:
147…152 °C
:
228-474-6
:
GHS07
:
Ambient
msds_ALFAA15415
Thermo Scientific Acroseal 25 Years