
p-Xylene-2-sulphonic acid hydrate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:609-54-1
:
Formula:C₈H₁₀O₃S
MDL Number:MFCD00064858
:186.23 g/mol
Melting Pt:84…86 °C
EINECS:210-193-5
:GHS07
Storage Temperature:Ambient