Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
alpha-Chloralose
ɑ-Chloralose
ɑ-Chloralose
ɑ-Chloralose
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
ɑ-Chloralose
Supplier:
Thermo Fisher Scientific
About this item
CAS Number:
15879-93-3
:
Formula:
C₈H₁₁Cl₃O₆
MDL Number:
MFCD00005542
:
309.52 g/mol
Related Information
Documentation
SDS
msds_ALFAA11492
Application Notes
Organic Building Blocks and Intermediates
Documentation