Zinc molybdate ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98% CAS No.: 13767-32-3 Molecular Formula: ZnMoO4 Formula Weight: 225.32 Physical Form: Powder Melting Point: 700°C MDL No.: MFCD00049622
Synonyms: Zinc molybdenum oxide
- CAS Number:13767-32-3
- Formula:MoO₄Zn
- MDL Number:MFCD00049622
- Molecular Weight:225.326 g/mol
- Melting Pt:>700 °C
- Storage Temperature:Ambient
- EINECS:237-377-8
2 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 13767-32-3 Molecular Formula: ZnMoO4 Formula Weight: 225.32 Physical Form: Powder Melting Point: 700°C MDL No.: MFCD00049622
Synonyms: Zinc molybdenum oxide
- CAS Number:13767-32-3
- Formula:MoO₄Zn
- MDL Number:MFCD00049622
- Molecular Weight:225.326 g/mol
- Melting Pt:>700 °C
- Storage Temperature:Ambient
- EINECS:237-377-8
Documents
Product Family Options
Product Information
- PkAvailabilityPrice



