

Ruthenium(III) chloride Ru ≥ 47.7%, anhydrous
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10049-08-8
:
Formula:Cl₃Ru
MDL Number:MFCD00011208
:207.43 g/mol
Merck Index:14,8302
Melting Pt:>500 °C (dec.)
Density:3.110
Storage Temperature:Ambient
EINECS:233-167-5
UN:3260
ADR:8,III
:GHS05
:GHS07