Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Polyethylene glycol methyl ether acrylate
Polyethylene glycol methyl ether acrylate, M.W. 1,000
Polyethylene glycol methyl ether acrylate, M.W. 1,000
Polyethylene glycol methyl ether acrylate, M.W. 1,000
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Polyethylene glycol methyl ether acrylate, M.W. 1,000
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
32171-39-4
:
Formula:
CH₃(OCH₂CH₂)nOCOC=CH₂
MDL Number:
MFCD00803685
Storage Temperature:
Refrigerator
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFA046537
Videos
Thermo Scientific Acroseal 25 Years
Documentation