Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Polyethylene glycol diacrylate (PEGDA)
Polyethylene glycol diacrylate (PEGDA), M.W. 1,000
Polyethylene glycol diacrylate (PEGDA), M.W. 1,000
Corrosive
Irritant
Polyethylene glycol diacrylate (PEGDA), M.W. 1,000
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Corrosive
Irritant
Polyethylene glycol diacrylate (PEGDA), M.W. 1,000
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
26570-48-9
:
Formula:
CH₂=COC(OCH₂CH₂)nOCOC=CH₂
MDL Number:
MFCD00081876
EINECS:
607-960-5
:
GHS05
:
GHS07
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFA046493
Videos
Thermo Scientific Acroseal 25 Years
Documentation