Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Poly(3-hexylthiophene-2,5-diyl)
Poly(3-hexylthiophene-2,5-diyl) regiorandom
Poly(3-hexylthiophene-2,5-diyl) regiorandom
Irritant
Poly(3-hexylthiophene-2,5-diyl) regiorandom
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
Poly(3-hexylthiophene-2,5-diyl) regiorandom
Supplier:
Thermo Fisher Scientific
About this item
CAS Number:
104934-50-1
:
110134-47-9
MDL Number:
MFCD00217686
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
SDS
msds_ALFA045906
Videos
Thermo Scientific Acroseal 25 Years
Documentation