Order Entry
Export
ContactUsLinkComponent
 
 
 
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), gauze woven from wire 500 mesh woven from 0.02mm (0.0009in) Type 304
  45140.RF
 :  
 
 
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), gauze woven from wire 500 mesh woven from 0.02mm (0.0009in) Type 304
  45140.RF
 :  

 

  • Pk:
    1
  • Seq:
    7780
  • Pk Info:
    30×30 cm

 

 

 :
 :55.845 g/mol
 :GHS08
 :GHS07
 :Ambient
 :Fe
 :MFCD00148499