

2-Methyl-1-pyrroline
Catalog # 44853.AC
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:25 mL
Specifications
About this item
:
:83.13 g/mol
Boiling Pt:104…105 °C
Density:0.878
Flash Pt:10 °C (50 °F)
EINECS:212-822-9
ADR:3,II
:GHS02
:GHS05
Storage Temperature:Ambient
CAS Number:872-32-2
Formula:C₅H₉N
MDL Number:MFCD00128806