0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Palladium nickel
Palladium nickel, gauze woven from wire 50 mesh woven from 0.1578mm (0.0062in)
Palladium nickel, gauze woven from wire 50 mesh woven from 0.1578mm (0.0062in)
:
Thermo Fisher Scientific
Palladium nickel, gauze woven from wire 50 mesh woven from 0.1578mm (0.0062in)
:
Thermo Fisher Scientific
:
:
Pd:Ni;₉₅:₅wt%
:
MFCD05863544
:
GHS08
:
GHS07
:
Ambient
msds_ALFA044698
Thermo Scientific Acroseal 25 Years