Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Fullerene buckytube/nanotube
Fullerene buckytube/nanotube single-walled >60%
Fullerene buckytube/nanotube single-walled >60%
44501.02
:
Thermo Fisher Scientific
:
Fullerene buckytube/nanotube single-walled >60%
44501.02
:
Thermo Fisher Scientific
:
Pk:
0,5 g
Seq:
3035
:
:
Ambient
:
1325
:
4.1,III
:
GHS02
:
GHS07
:
308068-56-6
:
MFCD00282904