
2-Methyl-4-trimethylsilyl-1-buten-3-yne ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:18387-60-5
:
Formula:(CH₃)₃SiC ΞCC(CH₃)=CH₂
MDL Number:MFCD00190206
:138.28 g/mol
Boiling Pt:32 °C (15 mmHg)
Density:0.778
Storage Temperature:Ambient
ADR:3,III
:GHS02