Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Boron nitride
Boron nitride 31% refractory brushable paint
Boron nitride 31% refractory brushable paint
Irritant
Boron nitride 31% refractory brushable paint
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
Boron nitride 31% refractory brushable paint
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
10043-11-5
:
Formula:
BN
MDL Number:
MFCD00011317
:
24.82 g/mol
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFA044086
Videos
Thermo Scientific Acroseal 25 Years
Documentation