Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Poly(methyl methacrylate)
Poly(methyl methacrylate) 600 micron
Poly(methyl methacrylate) 600 micron
Poly(methyl methacrylate) 600 micron
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Poly(methyl methacrylate) 600 micron
Supplier:
Thermo Fisher Scientific
About this item
CAS Number:
9011-14-7
:
Formula:
C₅H₈O₂([-CH₂C(CH₃)(CO₂CH₃)-]n)
MDL Number:
MFCD00134349
Storage Temperature:
Ambient
Related Information
Documentation
SDS
msds_ALFA043969
Videos
Thermo Scientific Acroseal 25 Years
Documentation