Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Polyethylene (CAS 9002-88-4)
Polyethylene UHMW
Polyethylene UHMW
Polyethylene UHMW
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Polyethylene UHMW
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
9002-88-4
:
Formula:
(CH₂CH₂)n
MDL Number:
MFCD00084423
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFA043951
Videos
Thermo Scientific Acroseal 25 Years
Documentation