



Potassium-tert-pentyloxide 14 - 18% (w/v) in cyclohexane
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:41233-93-6
:
Formula:C₅H₁₁KO
MDL Number:MFCD00064808
:126.24 g/mol
Density:0.80
Flash Pt:–18 °C (0 °F)
Storage Temperature:Ambient
EINECS:255-272-5
UN:2924
ADR:3,II
:GHS02
:GHS08
:GHS05
:GHS07