

Nickel iron ≥99.95% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 3.18 mm (0.125 in)
Catalog # 40888.KS
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
:
Formula:Ni:Fe;₈₁:₁₉%
MDL Number:MFCD01741928
:GHS08
:GHS07
Storage Temperature:Ambient