
Triphenylphosphine-3,3',3''-trisulphonic acid trisodium salt ≥85%, tech.
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:63995-70-0
:
Formula:C₁₈H₁₂Na₃O₉PS₃
MDL Number:MFCD00145472
:568.408 g/mol
EINECS:264-596-6
:GHS07
Storage Temperature:Ambient