Bismuth (III) phosphate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10049-01-1
:
Formula:BiO₄P
MDL Number:MFCD00049721
:303.95 g/mol
Merck Index:14,1275
Melting Pt: Dec.
Density:6.32
Storage Temperature:Ambient
EINECS:233-161-2