Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Platinum
Platinum 5% on alumina, paste 5R94
Platinum 5% on alumina, paste 5R94
Platinum 5% on alumina, paste 5R94
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Platinum 5% on alumina, paste 5R94
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
7440-06-4
:
Formula:
Pt
MDL Number:
MFCD00011179
:
195.08 g/mol
Storage Temperature:
Ambient
EINECS:
231-116-1
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFA038318
Videos
Thermo Scientific Acroseal 25 Years
Documentation