Lead(II)-2-ethylhexanoate
Supplier: Thermo Fisher Scientific
Jump to:
About this item
CAS No.: 301-08-6 Molecular Formula: C16H30O4Pb Formula Weight: 493.61 Physical Form: Viscous Liquid Density: 1.56 Odour: Odourless UN No.: UN3082 MDL No.: MFCD00014003
Synonyms: Lead(II) 2-ethylhexanoate
- CAS Number:301-08-6
- Formula:C₁₆H₃₀O₄Pb
- MDL Number:MFCD00014003
- Molecular Weight:493.61 g/mol
- Density:1.56
- Storage Temperature:Ambient
- EINECS:206-107-0
- ADR:9,III
2 Options Available Below
Product Details & Documents
CAS No.: 301-08-6 Molecular Formula: C16H30O4Pb Formula Weight: 493.61 Physical Form: Viscous Liquid Density: 1.56 Odour: Odourless UN No.: UN3082 MDL No.: MFCD00014003
Synonyms: Lead(II) 2-ethylhexanoate
- CAS Number:301-08-6
- Formula:C₁₆H₃₀O₄Pb
- MDL Number:MFCD00014003
- Molecular Weight:493.61 g/mol
- Density:1.56
- Storage Temperature:Ambient
- EINECS:206-107-0
- ADR:9,III



