Order Entry
Export
ContactUsLinkComponent
Mineral oil, white (petroleum)
Mineral oil, white (petroleum)
Catalog # 31911.D9
CAS Number:  
Mineral oil, white (petroleum)
Catalog # 31911.D9
CAS Number:  

Specifications

  • Pk:
    5
  • Seq:
    4032
  • Pk Info:
    5x1 kg

Specifications

About this item

 :
Flash Pt:>100 °C (212 °F)
Storage Temperature:Ambient
CAS Number:8042-47-5
MDL Number:MFCD00131611