Palladium 1% on alumina, powder, reduced
Catalog # 11711.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:106.420 g/mol
Storage Temperature:Ambient
CAS Number:7440-05-3
Formula:Pd
MDL Number:MFCD00011167