Order Entry
Export
ContactUsLinkComponent
Stainless steel (Fe:Cr:Ni; 70:17:13 wt%), powder -140 mesh Type 303-L
Health
Irritant
Stainless steel (Fe:Cr:Ni; 70:17:13 wt%), powder -140 mesh Type 303-L
Catalog # 11084.A1
CAS Number:  
Health
Irritant
Stainless steel (Fe:Cr:Ni; 70:17:13 wt%), powder -140 mesh Type 303-L
Catalog # 11084.A1
CAS Number:  

Specifications

  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :55.845 g/mol
 :GHS08
 :GHS07
Storage Temperature:Ambient
Formula:Fe
MDL Number:MFCD00148499