Anti-INHA Goat Polyclonal Antibody
Catalog # EVBIEB12851
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen name:inhibin alpha
- Antigen symbol:INHA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Format:affinity purified
- Form:liquid
- Antigen synonyms:AW555078|inhibin alpha chain
- Storage buffer:Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA.
- Sequence:C-QRPPEEPAAH
- Storage temperature:Store at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing
- Concentration:100µg/200µl
- Shipping temperature:8 °C
- Purification:Affinity purified
- Size:100µg
- Pack type:plastic vial
- Pk:0,1 mg
Specifications
About this item
Western Blot: Preliminary experiments gave an approx 30kDa band in Mouse Brain lysates after 2µg/ml antibody staining. Please note that currently we cannot find an explanation in the literature for the band we observe given the calculated size of 39.5kDa
Type: Primary
Antigen: INHA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat