Anti-VASP Goat Polyclonal Antibody
Catalog # EVBIEB12619
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen name:Vasodilator-stimulated phosphoprotein
- Antigen symbol:VASP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Bovine,Dog,Mouse
- Western blot:Yes
- Format:affinity purified
- Form:liquid
- Gene ID:7408
- Storage buffer:Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA.
- Sequence:C-RKVSKQEEASGGPT
- Storage temperature:Store at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing
- Concentration:100µg/200µl
- Shipping temperature:8 °C
- Purification:Affinity purified
- Size:100µg
- Pack type:plastic vial
- Pk:0,1 mg
Specifications
About this item
Western Blot: Approx 50kDa band observed in Human Platelets lysates (calculated MW of 39.8kDa according to NP_003361.1). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Reinhard et al, EMBO J. 1992 J
Type: Primary
Antigen: VASP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Dog, Bovine