Order Entry
Export
ContactUsLinkComponent
Anti-NNMT Goat Polyclonal Antibody
Anti-NNMT Goat Polyclonal Antibody
  EVBIEB12577
undefined
Anti-NNMT Goat Polyclonal Antibody
  EVBIEB12577

 

  • Antibody type:
    Primary
  • Antigen name:
    Nicotinamide N-methyltransferase
  • Antigen symbol:
    NNMT
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Format:
    affinity purified
  • Form:
    liquid
  • Gene ID:
    4837
  • Storage buffer:
    Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA.
  • Sequence:
    PDLPTYCRALRN
  • Storage temperature:
    Store at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing
  • Concentration:
    100µg/200µl
  • Shipping temperature:
    8 °C
  • Purification:
    Affinity purified
  • Size:
    100µg
  • Pack type:
    plastic vial
  • Pk:
    0,1 mg

 

 

Western Blot: Approx 28kDa band observed in lysates of cell line HeLa (calculated MW of 29..6kDa according to NP_006160.1). Recommended concentration: 0.3-1µg/ml.

Type: Primary
Antigen: NNMT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human