Anti-OLR1 Goat Polyclonal Antibody
EVBIEB11693
- Antibody type:Primary
- Antigen name:oxidized low density lipoprotein (lectin-like) receptor 1
- Antigen symbol:OLR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Format:affinity purified
- Form:liquid
- Gene ID:4973
- Antigen synonyms:SCARE1|CLEC8A|LOX1|SLOX1|LOXIN
- Storage buffer:Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA.
- Sequence:DLKIQTVKDQPDEK-C
- Storage temperature:Store at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing
- Concentration:100µg/200µl
- Shipping temperature:8 °C
- Purification:Affinity purified
- Size:100µg
- Pack type:plastic vial
- Pk:0,1 mg
Western Blot: Approx 40kDa band observed in Human Liver lysates (calculated MW of 30.9kDa according to NP_002534.1). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Yoshida et al, Biochem J. 1998 Aug
Type: Primary
Antigen: OLR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human