Anti-SP1 Goat Polyclonal Antibody
EVBIEB09068
- Antibody type:Primary
- Antigen name:trans-acting transcription factor 1
- Antigen symbol:SP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Pig,Rat,Mouse
- Western blot:Yes
- Format:affinity purified
- Form:liquid
- Antigen synonyms:Sp1-1|AI845540|AA450830|1110003E12Rik
- Storage buffer:Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA.
- Sequence:C-DGIHDETAGGEGEN
- Storage temperature:Store at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing
- Concentration:100µg/200µl
- Shipping temperature:8 °C
- Purification:Affinity purified
- Size:100µg
- Pack type:plastic vial
- Pk:0,1 mg
Western Blot: Approx 100kDa band observed in nuclear lysates of cell line NIH3T3 (calculated MW of 80.4kDa according to NP_038700.2). The observed molecular weight corresponds to earlier findings with different antibodies from other commercial sources. R
Type: Primary
Antigen: SP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat, Pig