Order Entry
Export
ContactUsLinkComponent
Anti-STAU1 Rabbit Polyclonal Antibody
Anti-STAU1 Rabbit Polyclonal Antibody
Catalog # BSBTPB9895
Supplier:  Boster Bio
undefined
Anti-STAU1 Rabbit Polyclonal Antibody
Catalog # BSBTPB9895
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Staufen double-stranded RNA binding protein 1
  • Antigen symbol:
    STAU1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    6780
  • Antigen synonyms:
    PPP1R150|STAU
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: STAU1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human