Order Entry
Export
ContactUsLinkComponent
Anti-RAP1A Rabbit Polyclonal Antibody
Anti-RAP1A Rabbit Polyclonal Antibody
Catalog # BSBTPB9816
Supplier:  Boster Bio
undefined
Anti-RAP1A Rabbit Polyclonal Antibody
Catalog # BSBTPB9816
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    RAP1A, Member Of RAS Oncogene Family
  • Antigen symbol:
    RAP1A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    5906
  • Antigen synonyms:
    KREV1|G-22K|RAP1|C21KG|KREV-1|SMGP21
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: RAP1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat