Order Entry
Export
Orders LinkContactUsLinkComponent
Anti-EIF2AK2 Rabbit Polyclonal Antibody

Anti-EIF2AK2 Rabbit Polyclonal Antibody

100 µg
Catalog # BSBTPB9803
Supplier:   Boster Bio
Anti-EIF2AK2 Rabbit Polyclonal Antibody
Catalog # BSBTPB9803
Supplier:  Boster Bio
Anti-EIF2AK2 Rabbit Polyclonal Antibody
Anti-EIF2AK2 Rabbit Polyclonal Antibody

100 µg

Price
546,00 €
Log in for availability
1 * 100 µG
Product Details & Documents

Type: Primary
Antigen: EIF2AK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Documents

Specifications
  • Size:
    100 µg
  • Western blot:
    Yes
  • Clonality:
    Polyclonal
  • Antibody type:
    Primary
  • Isotype:
    IgG
  • Host:
    Rabbit
  • Cross adsorption:
    No
  • Antigen name:
    Eukaryotic translation initiation factor 2-alpha kinase 2
  • Cat. No.:
    BSBTPB9803
  • Reactivity:
    Human
  • Gene ID:
    5610
  • Antigen synonyms:
    PPP1R83|PRKR|EIF2AK1|PKR
  • Conjugation:
    Unconjugated
  • Antigen symbol:
    EIF2AK2