Order Entry
Export
ContactUsLinkComponent
Anti-CD19 Rabbit Polyclonal Antibody
Anti-CD19 Rabbit Polyclonal Antibody
Catalog # BSBTPB9800
Supplier:  Boster Bio
undefined
Anti-CD19 Rabbit Polyclonal Antibody
Catalog # BSBTPB9800
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD19 molecule
  • Antigen symbol:
    CD19
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    930
  • Antigen synonyms:
    CVID3|B4
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: CD19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human