Order Entry
Export
ContactUsLinkComponent
Anti-TICAM1 Rabbit Polyclonal Antibody
Anti-TICAM1 Rabbit Polyclonal Antibody
  BSBTRP1100
undefined
Anti-TICAM1 Rabbit Polyclonal Antibody
  BSBTRP1100

 

  • Antibody type:
    Primary
  • Antigen name:
    Toll-like receptor adaptor molecule 1
  • Antigen symbol:
    TICAM1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    106759
  • Antigen synonyms:
    AW547018|AW046014|TRIF|TICAM-1
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: TICAM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse