Anti-Tsc2 Rabbit Polyclonal Antibody
Catalog # ABGEAP3826A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen symbol:Tsc2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Mouse
- Western blot:Yes
- Modification:phospho-Ser1412
- Storage buffer:pH 7,4
- Molecular weight:202.071 kDa
- Sequence:KIDIGRLBPEAKVRSC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.3
- Shipping temperature:4 °C
- Immunogen:This mouse TSC2 Antibody is generated from rabbits immunized with a KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding S1412 of mouse TSC2.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: Tsc2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse