Order Entry
Export
ContactUsLinkComponent
Anti-CASP3 Rabbit Polyclonal Antibody
Anti-CASP3 Rabbit Polyclonal Antibody
  ABGEAP3778D
 :  Abgent
Anti-CASP3 Rabbit Polyclonal Antibody
  ABGEAP3778D
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    caspase 3
  • Antigen symbol:
    CASP3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Gene ID:
    12367
  • Antigen synonyms:
    Lice|A830040C14Rik|Yama|AC-3|CASP-3|CPP32|CC3|mldy|SCA-1|CPP-32
  • Modification:
    phospho-Ser26
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    31.475 kDa
  • Sequence:
    KTIHGSKBVDSGIYC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.75
  • Shipping temperature:
    4 °C
  • Immunogen:
    This mouse CASP3 Antibody is generated from rabbits immunized with a KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding S26 of mouse CASP3.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: CASP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse