Anti-SEC11A Rabbit Polyclonal Antibody
Catalog # ABGEAP20099C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:SEC11 homolog A, signal peptidase complex subunit
- Antigen symbol:SEC11A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Gene ID:23478
- Antigen synonyms:SPCS4A|SEC11L1|1810012E07Rik|SPC18|sid2895
- Amino acid number:95 to 123
- Storage buffer:pH 7,4
- Molecular weight:20.625 kDa
- Sequence:KFLTKGDNNAVDDRC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.96
- Shipping temperature:4 °C
- Immunogen:This SC11A antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 95-123 amino acids from the Central region of human SC11A.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: SEC11A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human