Order Entry
Export
ContactUsLinkComponent
 
Anti-TMOD4 Rabbit Polyclonal Antibody
  ABGEAP17611C
 :  Abgent
Anti-TMOD4 Rabbit Polyclonal Antibody
  ABGEAP17611C
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    tropomodulin 4 (muscle)
  • Antigen symbol:
    TMOD4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    internal
  • Gene ID:
    29765
  • Antigen synonyms:
    SK-TMOD
  • Amino acid number:
    150 to 176
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    39.335 kDa
  • Sequence:
    VQPDKYKPVPDEC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    1.000
  • Shipping temperature:
    4 °C
  • Immunogen:
    This TMOD4 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 150-176 amino acids from the Central region of human TMOD4.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: TMOD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human