Order Entry
Export
ContactUsLinkComponent
Anti-RLIM Rabbit Polyclonal Antibody
Anti-RLIM Rabbit Polyclonal Antibody
  ABGEAP20050C
 :  Abgent
Anti-RLIM Rabbit Polyclonal Antibody
  ABGEAP20050C
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Ring Finger Protein, Lim Domain Interacting
  • Antigen symbol:
    RLIM
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    internal
  • Gene ID:
    51132
  • Antigen synonyms:
    RNF12|NY-REN-43
  • Amino acid number:
    199 to 227
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    68.549 kDa
  • Sequence:
    RSPDHRRTRARAERC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.56
  • Shipping temperature:
    4 °C
  • Immunogen:
    This RLIM antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 199-227 amino acids from the Central region of human RLIM.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: RLIM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human