Order Entry
Export
ContactUsLinkComponent
Anti-TIGAR Rabbit Polyclonal Antibody
Anti-TIGAR Rabbit Polyclonal Antibody
Catalog # ABGEAP19202A
Supplier:  Abgent
Anti-TIGAR Rabbit Polyclonal Antibody
Catalog # ABGEAP19202A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    TP53 induced glycolysis regulatory phosphatase
  • Antigen symbol:
    TIGAR
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Gene ID:
    57103
  • Antigen synonyms:
    C12orf5|FR2BP
  • Amino acid number:
    1 to 30
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    30.063 kDa
  • Sequence:
    VRHGETRFNKEKC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.35
  • Shipping temperature:
    4 °C
  • Immunogen:
    This TIGAR antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human TIGAR.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: TIGAR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Mouse