Order Entry
Export
ContactUsLinkComponent
Calcium carbonate 98.5-100.5% USP, FCC, Macron Fine Chemicals™
Calcium carbonate 98.5-100.5% USP, FCC, Macron Fine Chemicals™
Catalog # 4052-12
CAS Number:  
undefined
Calcium carbonate 98.5-100.5% USP, FCC, Macron Fine Chemicals™
Catalog # 4052-12
CAS Number:  

Specifications

  • Pack type:
    Poly bottle, round
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • USP - Assay (CaCO₃):
    98.0 - 100.5 %
  • USP - Identification:
    Passes Test
  • USP - Loss on Drying:
    = 2.0 %
  • USP - Acid-Insoluble Substances:
    = 0.2 %
  • USP - Barium (Ba):
    Passes Test
  • USP - Heavy Metals (as Pb):
    = 0.002 %
  • USP - Magnesium and Alkali Salts:
    = 1.0 %
  • USP - Limit of Fluoride:
    = 0.005 %
  • USP - Iron (Fe):
    = 0.1 %
  • USP - Mercury (Hg):
    = 0.5 ppm
  • FCC - Assay (CaCO₃):
    98.0 - 100.5 %
  • FCC - Identification:
    Passes Test
  • FCC - Loss on Drying:
    = 2.0 %
  • FCC - Acid-Insoluble Substances:
    = 0.2 %
  • FCC - Lead (Pb):
    = 3 mg/kg
  • FCC - Arsenic (As):
    = 0.0003 %
  • FCC - Magnesium and Alkali Salts:
    = 1.0 %
  • FCC - Limit of Fluoride:
    = 0.005 %

Specification Test Results

About this item

 :
 :100.09 g/mol
Merck Index:13,01658
Melting Pt:825 °C
Density:2.71 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:207-439-9
CAS Number:471-34-1
Formula:CaCO₃
MDL Number:MFCD00010906