Anti-V-ATPase, B Rabbit Polyclonal Antibody
Catalog # AGRIAS142775
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen name:V-ATPase, B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Arabidopsis,Vigna radiata,A. thaliana,N. tabacum,Halleri,Thellungiella salsuginea
- Western blot:Yes
- Immunogen:KLH-conjugated synthetic peptide chosen from Arabidopsis thaliana V-ATPase subunit B, isoform B1: UniProt: Q683E8, TAIR: AT1G76030, isoform B2 UniProt: Q9SZN1, TAIR: AT4G38510, isoform B3: UniProt: Q8W4E2 , TAIR: AT1G20260
- Size:50 µg
- Pk:50 µG
Specifications
About this item
V-ATPase, subunit B is a non-catalytic subunit of the peripheral V1 complex of vacuolar ATPase. Alternative names: Vacuolar proton pump subunit B1, vacuolar H+-ATPase subunit B, V-ATPase 57 kDa subunit.,V-ATPase, subunit B is a non-catalytic subunit of the peripheral V1 complex of vacuolar ATPase. Alternative names: Vacuolar proton pump subunit B1, vacuolar H+-ATPase subunit B, V-ATPase 57 kDa subunit.
Type: Primary
Antigen: V-ATPase, B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Arabidopsis, Halleri, A. thaliana, N. tabacum, Thellungiella salsuginea, Vigna radiata