Anti-LHY Rabbit Polyclonal Antibody
50 µg
Catalog # AGRIAS132661
Supplier: Agrisera
Jump to:
Product Details & Documents
LHY (Late elongated hypocotyl) is a transcription factor which is involved in the circadian clock. It binds to the promoter region of APRR1/TOC1 and TCP21/CHE to repress their transcription. Alternative names: LATE ELONGATED HYPOCOTYL, LATE ELONGATED HYPOCOTYL 1, LHY, LHY1.,LHY (Late elongated hypocotyl) is a transcription factor which is involved in the circadian clock. It binds to the promoter region of APRR1/TOC1 and TCP21/CHE to repress their transcription. Alternative names: LATE ELONGATED HYPOCOTYL, LATE ELONGATED HYPOCOTYL 1, LHY, LHY1.
Type: Primary
Antigen: LHY
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chlamydomonas reinhardtii
Documents
AS132661
Datasheet
Specifications
- Size:50 µg
- Western blot:Yes
- Cat. No.:AGRIAS132661
- Clonality:Polyclonal
- Reactivity:Chlamydomonas reinhardtii
- Antibody type:Primary
- Immunogen:KLH-conjugated synthetic peptide derived from Arabidopsis thaliana LHY protein sequence, UniProt: Q6R0H1, TAIR: AT1G01060
- Host:Rabbit
- Conjugation:Unconjugated
- Antigen symbol:LHY



