Anti-Rad51C Rabbit Polyclonal Antibody
Catalog # BSBTPB9792
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:RAD51 paralog C
- Antigen symbol:RAD51C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:5889
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for DNA repair protein RAD51 homolog 3(RAD51C) detection. Tested with WB in Human.
Type: Primary
Antigen: Rad51C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human